N-(5-Amino-2-fluorophenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048881-2.5G 048881-500MG
101885-432EA 364.62 USD
101885-432 101885-434
N-(5-Amino-2-fluorophenyl)-2-methoxyacetamide
N-(5-Amino-2-fluorophenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₉H₁₁FN₂O₂
MDL Number: MFCD09046349
CAS Number: 263400-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR