N-(3-Amino-4-methylphenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048880-500MG 048880-2.5G
101885-428EA 364.62 USD
101885-428 101885-430
N-(3-Amino-4-methylphenyl)-2-methoxyacetamide
N-(3-Amino-4-methylphenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD09939816
CAS Number: 946769-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR