N-(5-Amino-2-methylphenyl)-2-methoxyacetamide

Supplier: Matrix Scientific

048879-500MG 048879-2.5G
101885-424EA 364.62 USD
101885-424 101885-426
N-(5-Amino-2-methylphenyl)-2-methoxyacetamide
N-(5-Amino-2-methylphenyl)-2-methoxyacetamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD09049927
CAS Number: 926186-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR