N-(3-Amino-4-fluorophenyl)-2-furamide

Supplier: Matrix Scientific

048865-500MG 048865-2.5G
101885-368EA 364.62 USD
101885-368 101885-370
N-(3-Amino-4-fluorophenyl)-2-furamide
N-(3-Amino-4-fluorophenyl)-2-furamide

Hazard: Irritant

Formula: C₁₁H₉FN₂O₂
MDL Number: MFCD09049134
CAS Number: 926222-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR