N-(3-Amino-4-methylphenyl)isonicotinamide

Supplier: Matrix Scientific

048855-500MG 048855-2.5G
101885-328EA 364.62 USD
101885-328 101885-330
N-(3-Amino-4-methylphenyl)isonicotinamide
N-(3-Amino-4-methylphenyl)isonicotinamide

Hazard: Irritant

Formula: C₁₃H₁₃N₃O
MDL Number: MFCD02585703
CAS Number: 653584-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR