N-(4-Amino-2-methylphenyl)-2-ethoxyacetamide

Supplier: Matrix Scientific

048850-500MG 048850-2.5G
101885-310EA 838.46 USD
101885-310 101885-308
N-(4-Amino-2-methylphenyl)-2-ethoxyacetamide
N-(4-Amino-2-methylphenyl)-2-ethoxyacetamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD09731895
CAS Number: 953894-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR