N-(5-Amino-2-fluorophenyl)-2-ethoxyacetamide

Supplier: Matrix Scientific

048844-2.5G 048844-500MG
101885-284EA 364.62 USD
101885-284 101885-286
N-(5-Amino-2-fluorophenyl)-2-ethoxyacetamide
N-(5-Amino-2-fluorophenyl)-2-ethoxyacetamide

Hazard: Irritant

Formula: C₁₀H₁₃FN₂O₂
MDL Number: MFCD09730603
CAS Number: 953753-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR