[1-(4-Biphenylylmethyl)-3-oxo-2-piperazinyl]acetic acid

Supplier: Matrix Scientific

048832-500MG
101885-244EA 269.23 USD
101885-244
[1-(4-Biphenylylmethyl)-3-oxo-2-piperazinyl]acetic acid
[1-(4-Biphenylylmethyl)-3-oxo-2-piperazinyl]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₀N₂O₃
MDL Number: MFCD05666817
CAS Number: 1023919-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR