2-[1-(2-Ethoxybenzyl)-2-piperazinyl]ethanol

Supplier: Matrix Scientific

048831-500MG
101885-242EA 269.23 USD
101885-242
2-[1-(2-Ethoxybenzyl)-2-piperazinyl]ethanol
2-[1-(2-Ethoxybenzyl)-2-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₂
MDL Number: MFCD13248811
CAS Number: 1201633-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR