2-[1-(4-Ethoxybenzyl)-2-piperazinyl]ethanol

Supplier: Matrix Scientific

048829-500MG
101885-238EA 269.23 USD
101885-238
2-[1-(4-Ethoxybenzyl)-2-piperazinyl]ethanol
2-[1-(4-Ethoxybenzyl)-2-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₂
MDL Number: MFCD05666722
CAS Number: 671795-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR