2-[1-(3,5-Dimethoxybenzyl)-2-piperazinyl]ethanol

Supplier: Matrix Scientific

048825-500MG
101885-230EA 269.23 USD
101885-230
2-[1-(3,5-Dimethoxybenzyl)-2-piperazinyl]ethanol
2-[1-(3,5-Dimethoxybenzyl)-2-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₃
MDL Number: MFCD05666721
CAS Number: 671795-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR