2-[1-(1H-Indol-3-ylmethyl)-2-piperazinyl]ethanol

Supplier: Matrix Scientific

048823-500MG
101885-226EA 269.23 USD
101885-226
2-[1-(1H-Indol-3-ylmethyl)-2-piperazinyl]ethanol
2-[1-(1H-Indol-3-ylmethyl)-2-piperazinyl]ethanol

Hazard: Irritant

Formula: C₁₅H₂₁N₃O
MDL Number: MFCD05870669
CAS Number: 847480-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR