2-Methoxybutanoic acid
Supplier: Matrix Scientific
048813-500MG
101885-206EA
306.15
USD
101885-206
2-Methoxybutanoic acid
2-Methoxybutanoic acid
Hazard: Irritant
Formula:
C₅H₁₀O₃ |
MDL Number:
MFCD09864397 CAS Number: 56674-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...