6-Methoxyhexanoic acid

Supplier: Matrix Scientific

048812-500MG
101885-204EA 187.69 USD
101885-204
6-Methoxyhexanoic acid
6-Methoxyhexanoic acid

Hazard: Irritant

Formula: C₇H₁₄O₃
MDL Number: MFCD09759056
CAS Number: 41639-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR