N-Methyl-1-[5-(2-thienyl)-1H-pyrazol-3-yl]methanamine dihydrochloride

Supplier: Matrix Scientific

048809-500MG
101885-198EA 306.15 USD
101885-198
N-Methyl-1-[5-(2-thienyl)-1H-pyrazol-3-yl]methanamine dihydrochloride
N-Methyl-1-[5-(2-thienyl)-1H-pyrazol-3-yl]methanamine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂N₃S
MW: 266.19 g/mol
MDL Number: MFCD13248802
CAS Number: 1231953-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR