N,3,3-Trimethyl-2-butanamine hydrochloride

Supplier: Matrix Scientific

048806-500MG
101885-192EA 269.23 USD
101885-192
N,3,3-Trimethyl-2-butanamine hydrochloride
N,3,3-Trimethyl-2-butanamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₈ClN
MDL Number: MFCD09971468
CAS Number: 89979-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR