N3-(3-Hydroxypropyl)-β-alaninamide ethanedioate (salt)

Supplier: Matrix Scientific

048805-500MG
101885-190EA 269.23 USD
101885-190
N3-(3-Hydroxypropyl)-β-alaninamide ethanedioate (salt)
N3-(3-Hydroxypropyl)-beta-alaninamide ethanedioate (salt)

Hazard: Irritant

Formula: C₈H₁₆N₂O₆
MW: 236.23 g/mol
MDL Number: MFCD13248800
CAS Number: 1201633-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR