(5-Phenyl-1H-pyrazol-3-yl)methanol

Supplier: Matrix Scientific

048803-500MG
101885-186EA 306.15 USD
101885-186
(5-Phenyl-1H-pyrazol-3-yl)methanol
(5-Phenyl-1H-pyrazol-3-yl)methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD11046186
CAS Number: 179057-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR