1'H-Spiro[cyclopentane-1,2'-quinazolin]-4'(3'H)-one

Supplier: Matrix Scientific

048802-500MG
101885-184EA 306.15 USD
101885-184
1'H-Spiro[cyclopentane-1,2'-quinazolin]-4'(3'H)-one
1'H-Spiro[cyclopentane-1,2'-quinazolin]-4'(3'H)-one

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD00171145
CAS Number: 1135-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR