N,4-Dimethyl-2-pentanamine hydrochloride

Supplier: Matrix Scientific

048801-500MG
101885-182EA 269.23 USD
101885-182
N,4-Dimethyl-2-pentanamine hydrochloride
N,4-Dimethyl-2-pentanamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₈ClN
MDL Number: MFCD09971469
CAS Number: 89979-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR