N-Methyl-1-(2-naphthyl)ethanamine hydrochloride

Supplier: Matrix Scientific

048798-500MG
101885-176EA 233.85 USD
101885-176
N-Methyl-1-(2-naphthyl)ethanamine hydrochloride
N-Methyl-1-(2-naphthyl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆ClN
MDL Number: MFCD13195885
CAS Number: 1197565-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR