2-Ethoxyethyl-4-methylbenzenesulfonate

Supplier: Matrix Scientific

048792-500MG
101885-164EA 233.85 USD
101885-164
2-Ethoxyethyl-4-methylbenzenesulfonate
2-Ethoxyethyl-4-methylbenzenesulfonate

Hazard: Irritant

Formula: C₁₁H₁₆O₄S
MDL Number: MFCD00088078
CAS Number: 17178-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR