3-Ethoxypropyl-4-methylbenzenesulfonate
Supplier: Matrix Scientific
048791-500MG
101885-162EA
269.23
USD
101885-162
3-Ethoxypropyl-4-methylbenzenesulfonate
3-Ethoxypropyl-4-methylbenzenesulfonate
Hazard: Irritant
Formula:
C₁₂H₁₈O₄S |
MDL Number:
MFCD13248798 CAS Number: 91766-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...