3-Phenylpropyl-4-methylbenzenesulfonate

Supplier: Matrix Scientific

048790-500MG
101885-160EA 269.23 USD
101885-160
3-Phenylpropyl-4-methylbenzenesulfonate
3-Phenylpropyl-4-methylbenzenesulfonate

Hazard: Irritant

Formula: C₁₆H₁₈O₃S
MDL Number: MFCD00159241
CAS Number: 3742-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR