(1-Ethyl-3-oxo-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

048788-500MG
101885-156EA 269.23 USD
101885-156
(1-Ethyl-3-oxo-2-piperazinyl)acetic acid
(1-Ethyl-3-oxo-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₈H₁₄N₂O₃
MDL Number: MFCD08105445
CAS Number: 1048007-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR