Methyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 1-methylpyrrole-2-carboxylate
045456-500MG
045456-5G
045456-1G
101870-920EA
90.77
USD
101870-920
101870-918
101870-916
Methyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Hazard: Irritant
Formula:
C₇H₉NO₂ MW: 139.16 g/mol |
MDL Number:
MFCD00052747 CAS Number: 37619-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...