Methyl-3-amino-3-(2-thienyl)propanoate ≥95%

Supplier: Matrix Scientific

045384-1G
101870-576EA 698.46 USD
101870-576
Methyl-3-amino-3-(2-thienyl)propanoate ≥95%
Methyl-3-amino-3-(2-thienyl)propanoate

Hazard: Irritant

Formula: C₈H₁₁NO₂S
MW: 185.25 g/mol
MDL Number: MFCD03001286
CAS Number: 188812-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR