N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Methoxy-N-methyl-3-(trifluoromethyl)benzamide
045182-1G
045182-500MG
045182-5G
101869-560EA
90.77
USD
101869-560
101869-562
101869-558
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide ≥95%
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide
Hazard: Irritant
Formula:
C₁₀H₁₀F₃NO₂ MW: 233.19 g/mol |
MDL Number:
MFCD08689665 CAS Number: 116332-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...