2,4-Dibromo-3-isopropyl-6-methylbenzenol ≥95%

Supplier: Matrix Scientific

045099-500MG 045099-1G 045099-5G
101869-172EA 278.46 USD
101869-172 101869-174 101869-176
2,4-Dibromo-3-isopropyl-6-methylbenzenol ≥95%
2,4-Dibromo-3-isopropyl-6-methylbenzenol

Hazard: Irritant

Formula: C₁₀H₁₂Br₂O
MDL Number: MFCD02571954
CAS Number: 70454-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR