2-(1-Heptynyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

044911-1G 044911-5G 044911-500MG
101868-176EA 467.69 USD
101868-176 101868-174 101868-178
2-(1-Heptynyl)nicotinonitrile ≥95%
2-(1-Heptynyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₃H₁₄N₂
MDL Number: MFCD07366547
CAS Number: 885949-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR