4-Nitrobenzotriazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-1H-1,2,3-benzotriazole

044861-1G 044861-500MG
101867-930EA 150.77 USD
101867-930 101867-932
4-Nitrobenzotriazole ≥95%
4-Nitrobenzotriazole

Hazard: Irritant

Formula: C₆H₄N₄O₂
MW: 164.1216 g/mol
MDL Number: MFCD00160033
CAS Number: 6299-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR