Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate ≥95%
Supplier: Matrix Scientific
044832-1G
044832-500MG
044832-5G
101867-796EA
1109.23
USD
101867-796
101867-798
101867-800
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate ≥95%
Methyl-3,3-dimethyl-2-(1H-pyrrol-1-yl)butanoate
Hazard: Irritant
Formula:
C₁₁H₁₇NO₂ MW: 195.26 g/mol |
MDL Number:
MFCD01316055 CAS Number: 476335-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...