Methyl-3-phenyl-1,2,4-oxadiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
044711-1G
044711-500MG
101867-220EA
467.69
USD
101867-220
101867-218
Methyl-3-phenyl-1,2,4-oxadiazole-5-carboxylate ≥95%
Methyl-3-phenyl-1,2,4-oxadiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₀H₈N₂O₃ |
MDL Number:
MFCD08056635 CAS Number: 259150-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...