2-Methyl-2-morpholinopropanal ≥95%

Supplier: Matrix Scientific

044645-500MG 044645-5G 044645-1G
101866-884EA 595.38 USD
101866-884 101866-886 101866-882
2-Methyl-2-morpholinopropanal ≥95%
2-Methyl-2-morpholinopropanal

Hazard: Irritant/Refrigerate

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD05663791
CAS Number: 16042-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR