[3-Ethoxy-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide ≥95%
Supplier: Matrix Scientific
044543-1G
044543-500MG
101866-396EA
698.46
USD
101866-396
101866-394
[3-Ethoxy-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide ≥95%
[3-Ethoxy-2-(methoxyimino)-3-oxopropyl](triphenyl)phosphonium bromide
Hazard: Irritant
Formula:
C₂₄H₂₅BrNO₃P |
MDL Number:
MFCD11553020 CAS Number: 108411-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...