Methyl-2-(methylsulfinyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
044503-500MG
044503-5G
044503-1G
101866-212EA
307.69
USD
101866-212
101866-214
101866-216
Methyl-2-(methylsulfinyl)benzenecarboxylate ≥95%
Methyl-2-(methylsulfinyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀O₃S |
MDL Number:
MFCD03844851 CAS Number: 4850-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...