Potassium-4-[2-(2-thienyl)vinyl]-2-pyrimidinethiolate ≥95%

Supplier: Matrix Scientific

044488-1G 044488-5G 044488-500MG
101866-132EA 467.69 USD
101866-132 101866-134 101866-130
Potassium-4-[2-(2-thienyl)vinyl]-2-pyrimidinethiolate ≥95%
Potassium-4-[2-(2-thienyl)vinyl]-2-pyrimidinethiolate

Hazard: Irritant

Formula: C₁₀H₇KN₂S₂
MW: 258.41 g/mol
MDL Number: MFCD00663706
CAS Number: 318248-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR