4-Bromo-3-cyanoacetylindol

Supplier: Matrix Scientific

041128-500MG
101850-860EA 476.92 USD
101850-860
4-Bromo-3-cyanoacetylindol
4-Bromo-3-cyanoacetylindol

Hazard: Irritant

Formula: C₁₁H₇BrN₂O
MDL Number: MFCD11849915
CAS Number: 1171917-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR