1-Phenylsulfonyl-3-chloroacetylindole

Supplier: Matrix Scientific

041127-500MG
101850-858EA 606.15 USD
101850-858
1-Phenylsulfonyl-3-chloroacetylindole
1-Phenylsulfonyl-3-chloroacetylindole

Hazard: Irritant

Formula: C₁₆H₁₂ClNO₃S
MDL Number: MFCD11849927
CAS Number: 424789-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR