1-Boc-3-chloroacetylindole

Supplier: Matrix Scientific

041126-500MG
101850-856EA 590.77 USD
101850-856
1-Boc-3-chloroacetylindole
1-Boc-3-chloroacetylindole

Hazard: Irritant

Formula: C₁₅H₁₆ClNO₃
MDL Number: MFCD11849926
CAS Number: 916818-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR