1-(4-Methylamino-2-piperidin-3-yl-7,8-dihydro-5H-pyrido[4,3-d]pyrimidin-6-yl)ethanone diHCl

Supplier: Matrix Scientific

041116-250MG
101850-832EA 687.69 USD
101850-832
1-(4-Methylamino-2-piperidin-3-yl-7,8-dihydro-5H-pyrido[4,3-d]pyrimidin-6-yl)ethanone diHCl
1-(4-Methylamino-2-piperidin-3-yl-7,8-dihydro-5H-pyrido[4,3-d]pyrimidin-6-yl)ethanone diHCl

Hazard: Irritant

Formula: C₁₅H₂₅Cl₂N₅O
MDL Number: MFCD12028559
CAS Number: 1185295-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR