N-[1-(2-Aminoethyl)-1H-pyrazol-4-yl]benzamide dihydrochloride

Supplier: Matrix Scientific

041111-250MG
101850-822EA 558.46 USD
101850-822
N-[1-(2-Aminoethyl)-1H-pyrazol-4-yl]benzamide dihydrochloride
N-[1-(2-Aminoethyl)-1H-pyrazol-4-yl]benzamide dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆Cl₂N₄O
MW: 303.19 g/mol
MDL Number: MFCD12028554
CAS Number: 1185295-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR