2-(1-Phenethylpiperidin-4-yl)ethylamine

Supplier: Matrix Scientific

041109-250MG
101850-818EA 558.46 USD
101850-818
2-(1-Phenethylpiperidin-4-yl)ethylamine
2-(1-Phenethylpiperidin-4-yl)ethylamine

Hazard: Irritant

Formula: C₁₅H₂₄N₂
MDL Number: MFCD12028552
CAS Number: 132740-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR