3-(3-Aminomethylphenyl)-5-methyl-6,7-dihydro-5H-isoxazolo[4,5-c]pyridin-4-one hydrochloride
Supplier: Matrix Scientific
041107-250MG
101850-814EA
893.85
USD
101850-814
3-(3-Aminomethylphenyl)-5-methyl-6,7-dihydro-5H-isoxazolo[4,5-c]pyridin-4-one hydrochloride
3-(3-Aminomethylphenyl)-5-methyl-6,7-dihydro-5H-isoxazolo[4,5-c]pyridin-4-one hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₆ClN₃O₂ |
MDL Number:
MFCD12028551 CAS Number: 1185295-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...