Pyrrolidine-3-carboxylic acid (2-imidazol-1-ylethyl)amide dihydrochloride

Supplier: Matrix Scientific

041105-250MG
101850-810EA 558.46 USD
101850-810
Pyrrolidine-3-carboxylic acid (2-imidazol-1-ylethyl)amide dihydrochloride
Pyrrolidine-3-carboxylic acid (2-imidazol-1-ylethyl)amide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈Cl₂N₄O
MW: 281.19 g/mol
MDL Number: MFCD12028549
CAS Number: 1185304-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR