Methyl(6-methyl-2-piperidin-3-yl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-4-yl)amine diHCl
Supplier: Matrix Scientific
041102-250MG
101850-804EA
558.46
USD
101850-804
Methyl(6-methyl-2-piperidin-3-yl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-4-yl)amine diHCl
Methyl(6-methyl-2-piperidin-3-yl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-4-yl)amine diHCl
Hazard: Irritant
Formula:
C₁₄H₂₅Cl₂N₅ |
MDL Number:
MFCD12028546 CAS Number: 1185299-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...