3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid amide hydrochloride

Supplier: Matrix Scientific

041100-250MG
101850-800EA 893.85 USD
101850-800
3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid amide hydrochloride
3-Azepan-2-ylmethylbenzo[b]thiophene-2-carboxylic acid amide hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₁ClN₂OS
MDL Number: MFCD12028544
CAS Number: 1185299-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR