4-(4-Fluorophenyl)morpholine-2-carboxylic acid
Supplier: Matrix Scientific
041098-250MG
101850-796EA
893.85
USD
101850-796
4-(4-Fluorophenyl)morpholine-2-carboxylic acid
4-(4-Fluorophenyl)morpholine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₂FNO₃ |
MDL Number:
MFCD12028542 CAS Number: 1171917-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...