2-(Methylpiperidin-4-ylamino)ethanol dihydrochloride

Supplier: Matrix Scientific

041097-250MG
101850-794EA 558.46 USD
101850-794
2-(Methylpiperidin-4-ylamino)ethanol dihydrochloride
2-(Methylpiperidin-4-ylamino)ethanol dihydrochloride

Hazard: Irritant

Formula: C₈H₂₀Cl₂N₂O
MDL Number: MFCD09864394
CAS Number: 864655-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR