4-Piperidin-4-ylpyrimidine dihydrochloride

Supplier: Matrix Scientific

041095-250MG
101850-790EA 558.46 USD
101850-790
4-Piperidin-4-ylpyrimidine dihydrochloride
4-Piperidin-4-ylpyrimidine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₅Cl₂N₃
MW: 236.15 g/mol
MDL Number: MFCD12028540
CAS Number: 1185304-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR